MilliporeSigma
  • Final report on the safety assessment of Octyidodecyl Stearoyl Stearate.
Final report on the safety assessment of Octyidodecyl Stearoyl Stearate.
International journal of toxicology (2002-01-05)
R S Lanigan
ABSTRACT
Octyldodecyl Stearoyl Stearate functions as an occlusive skin-conditioning agent and as a nonaqueous viscosity-increasing agent in many cosmetic formulations. Current concentrations of use are between 0.7% and 23%, although historically higher concentrations were used. The chemical is formed by a high-temperature, acid-catalyzed esterification reaction of long-chain alcohols (primarily C-20) and a mixture of primarily C-18 fatty acids. Levels of stearic acid, octyldodecanol, and octylydocecyl hydroxystearate in the final product are 5% or less--no other residual compounds are reported. Only limited safety test data were available on Octyldodecyl Stearoyl Stearate, but previous safety assessments of long-chain alcohols and fatty acids found these precursors to be safe for use in cosmetic formulations. Octyldodecyl Stearoyl Stearate produced no adverse effects in acute exposures in rats. The chemical was mostly nonirritating to animal skin at concentrations ranging from 7.5% to 10%; one study did find moderate irritation in rabbit skin at a concentration of 7.5%. Clinical tests at a concentration of 10.4% confirmed the absence of significant irritation in humans. An ocular toxicity study in rabbits found no toxicity. No evidence of genotoxicity was found in either a mammalian test system or in the Ames test system, with or without metabolic activation. The available data on Octyldodecyl Stearoyl Stearate and the previously considered data on long-chain alcohols and fatty acids, however, did not provide a sufficient basis to make a determination of safety. Additional data needs include (1) chemical properties, including the octanol/water partition coefficient; and (2) if there is significant dermal absorption or if significant quantities of the ingredient may contact mucous membranes or be ingested, then reproductive and developmental toxicity data may be needed. Until such time as these data are received, the available data do not support the safety of Octyldodecyl Stearoyl Stearate as used in cosmetic formulations.
Product Description
Supelco
Octyldodecanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
2-Octyl-1-dodecanol, 97%