MilliporeSigma
  • Phenothiaziniums as putative photobactericidal agents for red blood cell concentrates.
Phenothiaziniums as putative photobactericidal agents for red blood cell concentrates.
Journal of chemotherapy (Florence, Italy) (2002-01-05)
M Wainwright, D A Phoenix, T E Smillie, D R Wareing
ABSTRACT
The antibacterial activities of Methylene Blue and several of its congeners were measured against Yersinia enterocolitica, a gram-negative pathogen known to exhibit significant growth at 4 degrees C and thus constituting a threat to red blood cell concentrates which are stored at this temperature. None of the derivatives was highly active in dark conditions, as expected, but on illumination using a lamp emitting light in the waveband 615-645 nm, considerable bactericidal activity was noted using similar photosensitizer concentrations to those used elsewhere to inactivate blood-borne viruses. Two novel compounds in this area, the phenothiazinium New Methylene Blue N and the phenoxazinium Brilliant Cresyl Blue, exhibited bactericidal activity at lower concentrations than both of the established phenothiaziniums, Methylene Blue and Toluidine Blue O and the recently published blood photovirucidal agent 1,9-Dimethyl Methylene Blue. The photoactivity of these compounds was undiminished in the presence of red blood cells.
Sigma-Aldrich
New Methylene Blue N, Dye content ~70 %