MilliporeSigma
  • 1,2-diacetylbenzene, the neurotoxic metabolite of a chromogenic aromatic solvent, induces proximal axonopathy.
1,2-diacetylbenzene, the neurotoxic metabolite of a chromogenic aromatic solvent, induces proximal axonopathy.
Toxicology and applied pharmacology (2001-12-13)
M S Kim, M I Sabri, V H Miller, R J Kayton, D A Dixon, P S Spencer
ABSTRACT
Several widely used aromatic hydrocarbon solvents reportedly induce blue-green discoloration of tissues and urine in animals and humans. The chomophore has been proposed to result from a ninhydrin-like reaction with amino groups in proteins. The present study examines the neurotoxic property of 1,2-diacetylbenzene (1,2-DAB), the active metabolite of the chromogenic and neurotoxic aromatic solvent 1,2-diethylbenzene. Rats treated with 1,2-DAB, but not with the nonchromogenic isomer 1,3-DAB or with ninhydrin developed blue discoloration of internal organs, including the brain and spinal cord. Only 1,2-DAB induced limb weakness associated with nerve fiber changes, which were most prominent in spinal cord and spinal roots. Changes began with the formation of proximal, neurofilament-filled axonal swellings of the type seen after treatment with 3,4-dimethyl-2,5-hexanedione, a potent derivative of the active metabolite of the neurotoxic aliphatic hydrocarbon solvents n-hexane and methyl n-butyl ketone. These compounds are metabolized to a gamma-diketone that forms pyrroles with target proteins, such as neurofilament proteins. A comparable mechanism is considered for 1,2-DAB, an aromatic gamma-diketone.
Sigma-Aldrich
1,2-Diethylbenzene, technical grade, 92%
Pricing and availability is not currently available.
Sigma-Aldrich
1,2-Diacetylbenzene, 99%
Pricing and availability is not currently available.
Sigma-Aldrich
1,2-Diethylbenzene, ≥99.0% (GC)
Pricing and availability is not currently available.