MilliporeSigma
  • Liquid chromatographic/mass spectrometric determination of desmedipham and phenmedipham and their metabolites in soil.
Liquid chromatographic/mass spectrometric determination of desmedipham and phenmedipham and their metabolites in soil.
Journal of AOAC International (2001-10-17)
D Perret, A Gentili, S Marchese, A Marin, F Bruno
ABSTRACT
A simple method is described for the simultaneous determination of residues of 2 carbamate herbicides (phenmedipham and desmedipham) and related metabolites (m-aminophenol, aniline, and m-toluidine) in soil. The analytes are extracted from spiked soils with methanol. The solvent/soil suspension is centrifuged, and the supernatant is directly injected, without any further cleanup, into a reversed-phase liquid chromatography/mass spectrometry apparatus equipped with a TurbolonSpray interface. The method was tested on 5 soils having different physicochemical properties. Recoveries from the soil types, spiked over the range of 50-200 ppb, were essentially quantitative for each analyte. The detection limits of the method are < or = 25 ng/g.
Product Description
Supelco
Desmedipham, PESTANAL®, analytical standard
Supelco
Phenmedipham, PESTANAL®, analytical standard