MilliporeSigma
  • Drug sensitivity of Candida yeast isolated from patients with allergic diseases.
Drug sensitivity of Candida yeast isolated from patients with allergic diseases.
Bulletin of experimental biology and medicine (2001-09-11)
V G Arzumanyan, B F Semenov
ABSTRACT
Viability of 40 Candida spp. cultures was studied after long-term exposure to antifungal drugs in minimum inhibitory concentrations. The fungicidal effect decreased in the series: pimafucin-nitrofungin-diflucan-orungal-levorine-clotrimazole-exoderil. Nizoral in a concentration of 4 microg/ml was ineffective; in the rest cultures the effect was either fungistatic (of different degree) or null. Pimafucin, diflucan, nitrofungin, orungal, levorine, and exoderil possessed individual fungicidal effects.
Sigma-Aldrich
2-Chloro-4-nitrophenol, 97%