MilliporeSigma
  • Layered double hydroxide stability. 2. Formation of Cr(III)-containing layered double hydroxides directly from solution.
Layered double hydroxide stability. 2. Formation of Cr(III)-containing layered double hydroxides directly from solution.
Chemistry of materials : a publication of the American Chemical Society (2001-09-07)
J W Boclair, P S Braterman, J Jiang, S Lou, F Yarberry
ABSTRACT
Solutions containing divalent metal [M(II) = Mg2+, Zn2+, Co2+, Ni2+, Mn2+] chlorides and CrCl3 6H2O were titrated with NaOH to yield, for M(II) = Zn, Co, and Ni, hydrotalcite-like layered double hydroxides (LDHs), [[M(II)]1-z[Cr(III)]z(OH)2][Cl]z yH2O, in a single step, without intermediate formation of chromium hydroxide. Analysis of the resultant titration curves yields solubility constants for these compounds. These are in the order Zn < Ni approximately Co, with a clear preference for formation of the phase with z = 1/3. With Mg2+ as chloride, titration gives a mixture of Cr(OH)3 and Mg(OH)2, but the metal sulfates give Mg2Cr(OH)6 1/2(SO4) by a two-step process. Titrimetric and spectroscopic evidence suggests short-range cation order in the one-step LDH systems.
Sigma-Aldrich
Chromium(III) chloride, anhydrous, 99.99% trace metals basis
Sigma-Aldrich
Chromium(III) chloride, purified by sublimation, 99%