MilliporeSigma
  • Mild and selective sodium azide mediated cleavage of p-nitrobenzoic esters.
Mild and selective sodium azide mediated cleavage of p-nitrobenzoic esters.
Organic letters (2001-08-03)
J A Gómez-Vidal, M T Forrester, R B Silverman
ABSTRACT
[reaction: see text] A mild and selective cleavage of p-nitrobenzoic esters by sodium azide in methanol is reported. This new methodology is mild enough for use with acid- or base-sensitive compounds. No elimination byproducts are formed. Fmoc- and trifluoroacetyl-amino protecting groups, benzyl esters, and ethyl esters remain unaffected. Less reactive compounds are discussed in terms of steric factors, and yields are increased by altering the azide solvation.
Sigma-Aldrich
4-Nitrobenzoic acid, 98%
Sigma-Aldrich
4-Nitrobenzoic acid, purum, ≥98.0% (HPLC)