MilliporeSigma
  • Combination therapy improves the recovery of the skin barrier function: an experimental model using a contact allergy patch test combined with TEWL measurements.
Combination therapy improves the recovery of the skin barrier function: an experimental model using a contact allergy patch test combined with TEWL measurements.
Dermatology (Basel, Switzerland) (2001-07-17)
J P Hachem, K De Paepe, E Vanpée, L Kaufman, V Rogiers, D Roseeuw
ABSTRACT
Nickel (Ni) allergic contact dermatitis (ACD) alters the skin barrier. Our aim was to compare the efficacy of combination therapies on ACD, using a topical corticosteroid and a corneotherapy agent (barrier cream), with that of a single therapy with corticosteroids. On day 1, 3 Ni test patches were applied on each forearm of 14 Ni-patch-test-positive females. Four contained 5% Ni and 2 physiological saline. Either topical corticosteroid or barrier cream were matched with the combination of both products on 3 of the 4 Ni ACD. The fourth was not treated. Clinical scoring, transepidermal water loss (TEWL) and stratum corneum (SC) capacitance were measured before (day 1) and after (days 4-8) ACD. The combination therapy showed a significant decrease in TEWL values and an increase in SC capacitance. Combining a topical corticosteroid with corneotherapy agents prevents the delay in the healing process of skin barrier disruption due to ACD.
Product Description
Supelco
Hexachlorophene, PESTANAL®, analytical standard