MilliporeSigma
  • Microbial resolution of 2-hydroxy-3-nitropropionic acid for synthesis of optically active isoserine.
Microbial resolution of 2-hydroxy-3-nitropropionic acid for synthesis of optically active isoserine.
Bioscience, biotechnology, and biochemistry (2001-07-07)
Y Yasohara, J Hasegawa
ABSTRACT
The biocatalytic stereoselective hydrolysis of 2-hydroxy-3-nitropropionic acid esters was studied. Forty enzymes and three hundred microorganism strains were examined for their ability to hydrolyze ethyl 2-hydroxy-3-nitropropionic acid. Nocardia globerula IFO13150 gave n-butyl (R)-2-hydroxy-3-nitropropionate with a 92% enantiomeric excess (ee) and the corresponding carboxylic acid with a 92%ee, which was easily converted to (S)-isoserine, a useful beta-amino acid.
Sigma-Aldrich
DL-Isoserine, 98%