MilliporeSigma
  • The copper-mediated cross-coupling of phenylboronic acids and N-hydroxyphthalimide at room temperature: synthesis of aryloxyamines.
The copper-mediated cross-coupling of phenylboronic acids and N-hydroxyphthalimide at room temperature: synthesis of aryloxyamines.
Organic letters (2001-06-30)
H M Petrassi, K B Sharpless, J W Kelly
ABSTRACT
[figure: see text] A novel route to aryloxyamines via the copper-mediated cross-coupling of N-hydroxyphthalimide and phenylboronic acids is reported. The reaction is mediated by selected copper(I) and (II) salts in the presence of pyridine and is tolerant of several functional groups on the phenylboronic acid. The phthallmide group is removed using hydrazine to afford the corresponding aryloxyamine.
Sigma-Aldrich
N-Hydroxyphthalimide, 97%
Pricing and availability is not currently available.