MilliporeSigma
  • Structural requirements of phenol derivatives for direct activation of chloride currents via GABA(A) receptors.
Structural requirements of phenol derivatives for direct activation of chloride currents via GABA(A) receptors.
European journal of pharmacology (2001-06-12)
B Mohammadi, G Haeseler, M Leuwer, R Dengler, K Krampfl, J Bufler
ABSTRACT
Propofol directly activates gamma-aminobutyric acid (GABA(A)) receptors in the absence of the natural agonist. This mechanism is supposed to contribute to its sedative-hypnotic actions. We studied the effects of seven structurally related phenol derivatives on chloride inward currents via rat alpha1beta2gamma2 GABA(A) receptors, heterologously expressed in HEK 293 cells in order to find structural determinants for this direct agonistic action. Only compounds with the phenolic hydroxyl attached directly to the benzene ring and with aliphatic substituents in ortho position to the phenolic hydroxyl activated chloride currents in the absence of GABA. Concentrations required for half-maximum effect were 980 microM for 2-methylphenol, 230 microM for 2,6-dimethylphenol, 200 microM for thymol, and 23 microM for propofol. Drug-induced chloride currents showed no desensitisation during the 2-s application. These results show that the position of the aliphatic substituents with respect to the phenolic hydroxyl group is the crucial structural feature for direct GABA(A) activation by phenol derivatives.
Sigma-Aldrich
2,6-Xylenol, ≥99%, FG
Sigma-Aldrich
2,6-Dimethylphenol, 99%
Sigma-Aldrich
2,6-Dimethylphenol, ≥99.5%
Supelco
2,6-Dimethylphenol, PESTANAL®, analytical standard