MilliporeSigma
  • Pharmacological properties of R-755, a novel acyl-CoA:cholesterol acyltransferase inhibitor, in cholesterol-fed rats, hamsters and rabbits.
Pharmacological properties of R-755, a novel acyl-CoA:cholesterol acyltransferase inhibitor, in cholesterol-fed rats, hamsters and rabbits.
Japanese journal of pharmacology (2001-06-05)
Y Matsui, K Horiuchi, K Yamamoto, K Kanai
ABSTRACT
R-755 (N-(2,6-diethylphenyl)-N'-[3-(2-methylphenyl)-6,7-dihydro-5H-cyclopenta[f[l]benzothiophen-2-yl]urea), a novel acyl-CoA:cholesterol acyltransferase (ACAT) inhibitor, has been characterized in vitro, ex vivo and in vivo. R-755 potently inhibited ACAT activities, with IC50 values from 2.5 to 64 nM, in rabbit intestinal microsomes and several cell lines (CaCo-2, THP-1 and J774A.1 cells). R-755 reduced serum cholesterol and triglyceride levels and liver cholesterol contents in cholesterol-fed rats, hamsters and rabbits. Rabbits were fed a high cholesterol diet for 2 weeks and further fed the same diet containing R-755 for 2 weeks. R-755 dose-dependently reduced cholesterol content and ACAT activity in the aorta. When phorbol 12-myristate 13-acetate-treated THP-1 and J774A.1 cells were incubated in the medium containing 20% of serum from rats administered R-755, the ACAT activities of the cells were inhibited. Rabbits were fed a high cholesterol diet for 8 weeks to establish aortic atherosclerosis and then fed a normal diet with or without R-755 for 8 weeks. R-755 dose-dependently reduced the surface area with atherosclerotic involvement and cholesterol contents in the aorta, although plasma cholesterol level did not differ from that in the control group. These results suggest that R-755 is a potent hypolipidemic agent and has a direct antiatherosclerotic activity at the arterial wall.
Sigma-Aldrich
CI 976, >98% (HPLC), solid