MilliporeSigma
  • Mitochondrial DNA depletion associated with partial complex II and IV deficiencies and 3-methylglutaconic aciduria.
Mitochondrial DNA depletion associated with partial complex II and IV deficiencies and 3-methylglutaconic aciduria.
Journal of child neurology (2001-04-09)
F Scaglia, V R Sutton, O A Bodamer, H Vogel, S K Shapira, R K Naviaux, G D Vladutiu
ABSTRACT
We report a patient with mitochondrial DNA depletion, partial complex II and IV deficiencies, and 3-methylglutaconic aciduria. Complex II deficiency has not been previously observed in mitochondrial DNA depletion syndromes. The observation of 3-methylglutaconic and 3-methylglutaric acidurias may be a useful indicator of a defect in respiratory chain function caused by mitochondrial DNA depletion.
Sigma-Aldrich
3-Methylglutaric acid, 99%
Sigma-Aldrich
3-Hydroxy-3-methylglutaric acid, ≥95%