MilliporeSigma
  • Spectrophotometric determination of ferbam (iron(III) dimethyldithiocarbamate) in commercial sample and wheat grains using 4,7-diphenyl-1,10-phenanthroline after extraction into mesityl oxide.
Spectrophotometric determination of ferbam (iron(III) dimethyldithiocarbamate) in commercial sample and wheat grains using 4,7-diphenyl-1,10-phenanthroline after extraction into mesityl oxide.
Journal of environmental monitoring : JEM (2001-03-17)
A K Malik
ABSTRACT
A procedure has been developed for the determination of iron(III) dimethyldithiocarbamate by converting it into iron(II)-bathophenanthroline complex, which is then extracted into mesityl oxide in the presence of potassium perchlorate. The absorbance is then measured at 534 nm against a reagent blank. Beer's law is obeyed over the concentration range 0.5-20 microg ml(-1) in the final solution. The method is sensitive, highly selective and can be used for the determination of ferbam in a commercial sample, and in mixtures with various dithiocarbamates (ziram, zineb, maneb, etc.) and in wheat grains.
Sigma-Aldrich
Mesityl oxide, technical grade, 90%