MilliporeSigma
Activity profile of glycolamide ester prodrugs of ibuprofen.
Drug development and industrial pharmacy (2001-03-15)
A K Bansal, R K Khar, R Dubey, A K Sharma
ABSTRACT
Glycolamide esters of ibuprofen (I), namely, unsubstituted (II), N,N dimethyl (III), and N,N diethyl (IV), were synthesized and studied for different physicochemical, pharmacological, and toxicological properties. They were comparable with I in respect of anti-inflammatory and analgesic activity but did not exhibit reduction in the ulcerogenicity on oral administration. However, all three exhibited significantly better topical activity in carrageenan-induced rat paw edema assay. In the same assay, they provided significant protection against inflammation when applied at a site remote to the inflammation site.
Sigma-Aldrich
Glycolamide, 98%