MilliporeSigma
  • Complexation of trivalent lanthanide cations by galactitol in the solid state. The crystal structure and an FT-IR study of 2NdCl3.galactitol.14H2O.
Complexation of trivalent lanthanide cations by galactitol in the solid state. The crystal structure and an FT-IR study of 2NdCl3.galactitol.14H2O.
Carbohydrate research (2001-02-24)
L Yang, Y Zhao, W Tian, X Jin, S Weng, J Wu
ABSTRACT
The crystal structure of 2NdCl3.galactitol.14H2O has been determined. The crystal system is triclinic, space group: -1, with unit-cell dimensions: a = 9.736(2), b = 10.396, c = 8.027 A; alpha = 108.05(3), beta = 92.68(3), gamma = 88.44(3) degrees, V= 771.6(3) A3, Z = 2. Each Nd atom is coordinated to nine oxygen atoms, three from the alditol and six from water molecules, with Nd-O distances from 2.461 to 2.552 A. The seventh water molecule is hydrogen-bonded by the hydroxyl hydrogen on O-1 (O-1-H-ll...O-10, 2.639 A). The FT-IR spectra of 2NdCl3.galactitol.14H2O and 2PrCl3.galactitol.14H2O are analogous, and show that Pr and Nd have the same coordination mode. The IR results are consistent with the crystal structures.
Sigma-Aldrich
Neodymium(III) chloride, anhydrous, powder, ≥99.99% trace metals basis