MilliporeSigma
  • Charge-transfer complex formation between o-chloranil and a series of polynuclear aromatic hydrocarbons.
Charge-transfer complex formation between o-chloranil and a series of polynuclear aromatic hydrocarbons.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2001-02-24)
B Chakraborty, A K Mukherjee, B K Seal
ABSTRACT
The equilibrium constants, enthalpies and entropies of formation of molecular electron donor-acceptor (EDA) complexes of o-chloranil with a series of aromatic hydrocarbons have been determined spectrophotometrically. Spectroscopic and thermodynamic aspects of these complexes have been analysed.
Sigma-Aldrich
3,4,5,6-Tetrachloro-1,2-benzoquinone, 97%