MilliporeSigma
  • Simultaneous determination of paracetamol and methocarbamol in tablets by ratio spectra derivative spectrophotometry and LC.
Simultaneous determination of paracetamol and methocarbamol in tablets by ratio spectra derivative spectrophotometry and LC.
Journal of pharmaceutical and biomedical analysis (2001-02-24)
N Erk, Y Ozkan, E Banoğlu, S A Ozkan, Z Sentürk
ABSTRACT
The application of the ratio spectra derivative spectrophotometry and high-performance liquid chromatography (HPLC) to the simultaneous determination of paracetamol (PAR) and methocarbamol (MET) in combined pharmaceutical tablets is presented. The spectrophotometric procedure is based on the use of the first derivative of the ratio spectra obtained by dividing the absorbtion spectrum of the binary mixtures by a standard spectrum of one of the compounds. The first derivative amplitudes were measured at 243.0 and 230.3 nm for the assay of PAR and MET, respectively. Calibration graphs were established for 2-30 microg ml for PAR and 2-36 microg/ml for MET in binary mixture. The detection limits for PAR and MET were found 0.097 and 0.079 microg/ml, respectively; while the quantification limits were 0.573 microg/ml for PAR and 1.717 microg/ml for MET. For the HPLC procedure, a reversed-phase column with a mobile phase of methanol-water (60:40, v/v), was used to separate both compounds with a detection of 274.0 nm. Linearity was obtained in the concentration range of 2 300 and 1.5-375 microg/ml for PAR and MET, respectively. The detection and quantification limits were found to be 0.42 and 1.4 microg/ml for PAR and 0.36 and 1.2 microg/ml for MET, respectively. The relative standard deviations were found to be less than 0.52%, indicating reasonable repeatibility of both methods. The proposed methods were successfully applied to the determination of these drugs in commercial tablets.
Product Description
Supelco
Methocarbamol, Pharmaceutical Secondary Standard; Certified Reference Material