MilliporeSigma
  • Synergic activity of D-cycloserine and beta-chloro-D-alanine against Mycobacterium tuberculosis.
Synergic activity of D-cycloserine and beta-chloro-D-alanine against Mycobacterium tuberculosis.
The Journal of antimicrobial chemotherapy (2001-02-07)
S David
ABSTRACT
D-Cycloserine (DCS) is a peptidoglycan inhibitor. Although very effective against Mycobacterium tuberculosis, it is seldom employed in the management of this infection due to its high toxicity. beta-Chloro-D-alanine (another peptidoglycan inhibitor) reduces the MIC of DCS from 50 to 2.5 mg/L at a concentration significantly below its MIC for this organism. A reduction in bacterial viability and significant growth inhibition (as quantified by the X/Y quotient for the Bactec radiometric procedure) were observed with subinhibitory concentrations of both drugs. It is suggested that this powerful synergic effect should be the object of in vivo and eventually clinical trials.
Product Description
Supelco
Cycloserine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
β-Chloro-L-alanine hydrochloride, Alanine aminotransferase inhibitor
Sigma-Aldrich
β-Chloro-D-alanine hydrochloride