MilliporeSigma
  • Catalytic activity of anion-exchange resins modified with metal-porphine in oxidative reactions of phenols.
Catalytic activity of anion-exchange resins modified with metal-porphine in oxidative reactions of phenols.
Chemical & pharmaceutical bulletin (2000-11-22)
A Iwado, M Mifune, J Kato, J Oda, M Chikuma, N Motohashi, Y Saito
ABSTRACT
Anion-exchange resins modified with metal-porphine (M-Pr) have been investigated to develop a solid catalyst in the oxidative reaction of phenols by O2 in air. Co-Pr, which is easily prepared and separable from the reaction mixture, has been proved to accelerate the oxidative reaction of phenols such as 3,5-di-tertbutyl-4-hydroxyanisole. The resulting main oxidative products were identified to be quinones by using the GC-MS method.
Sigma-Aldrich
2,4-Di-tert-butylphenol, 99%
Sigma-Aldrich
2,5-Di-tert-butyl-4-methoxyphenol, 97%