MilliporeSigma
  • Galabiosyl donors; efficient synthesis from 1,2,3,4, 6-penta-O-acetyl-beta-D-galactopyranose.
Galabiosyl donors; efficient synthesis from 1,2,3,4, 6-penta-O-acetyl-beta-D-galactopyranose.
Carbohydrate research (2000-11-22)
J Ohlsson, G Magnusson
ABSTRACT
1,2,3,4,6-Penta-O-acetyl-beta-D-galactopyranose was transformed into phenyl 2,3,4,6-tetra-O-benzyl-1-thio-beta-D-galactopyranoside (5) and 4-methoxyphenyl 2,3,6-tri-O-benzoyl-beta-D-galactopyranoside (8) in 73% (two steps) and 58% (three steps) yield, respectively. Glycosylation of the acceptor 8 with donor 5 using N-iodosuccinimide-trimethylsilyl trifluoromethanesulfonate as promoter furnished the galabioside 9 (8.8 g) in 95% yield. Further transformations provided in high yields anomerically-activated galabiosides (thioglycoside (1), trichloroacetimidate (2), and bromosugar (3)) suitable for use as glycosyl donors in syntheses of galabiose-containing oligosaccharides. Several of the compounds reported here are crystalline, which greatly simplified purifications.
Sigma-Aldrich
β-D-Galactose pentaacetate, 98%
Sigma-Aldrich
Digalacturonic acid, ≥85% (HPLC)