MilliporeSigma
  • Sorption and photolysis studies in soil and sediment of the herbicide napropamide.
Sorption and photolysis studies in soil and sediment of the herbicide napropamide.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes (2000-11-09)
J P Aguer, L Cox, C Richard, M C Hermosin, J Cornejo
ABSTRACT
The influence of soil and sediment composition on sorption and photodegradation of the herbicide napropamide [N,N-diethyl-2-(1-naphthyloxy)propionamide] was investigated. Five soils and one sediment were selected for this study and the clay fractions were obtained by sedimentation. Sorption-desorption was studied by batch equilibration technique and photolysis in a photoreactor emitting within 300-450 nm wavelength with a maximum at 365 nm. Sorption increased with clay content and was not related to organic matter content. High irreversibility of sorption was related to the greater montmorillonite content. The presence of soil or sediment reduced photolysis rate due to screen effect and this process did not depend on solid composition but on particle size distribution.
Product Description
Supelco
Napropamide, PESTANAL®, analytical standard