MilliporeSigma
  • Rapid and sensitive determination of 4-nitrophenol, 3-methyl-4-nitrophenol, 4,6-dinitro-o-cresol, parathion-methyl, fenitrothion, and parathion-ethyl by liquid chromatography with electrochemical detection.
Rapid and sensitive determination of 4-nitrophenol, 3-methyl-4-nitrophenol, 4,6-dinitro-o-cresol, parathion-methyl, fenitrothion, and parathion-ethyl by liquid chromatography with electrochemical detection.
Journal of agricultural and food chemistry (2000-10-29)
T Galeano-Díaz, A Guiberteau-Cabanillas, N Mora-Díez, P Parrilla-Vázquez, F Salinas-López
ABSTRACT
Liquid chromatography with electrochemical detection has been used to determine various nitropesticides, DNOC, fenitrothion, and parathion (methyl and ethyl), and some of their main metabolites, 4-nitrophenol for parathion (methyl and ethyl) and 3-methyl-4-nitrophenol for fenitrothion, by using indirect detection. Analysis of them in river water samples has been performed without a preconcentration step. The recovery efficiencies of the tested compounds yielded values between 96 and 112% at the fortification level of 0.5 ppb in a river water sample, and their relative standard deviations were between 1 and 15%. The detection limits of these compounds ranged between 0.05 and 0.14 ppb.
Product Description
Supelco
DNOC, PESTANAL®, analytical standard
Supelco
Parathion-methyl, PESTANAL®, analytical standard