MilliporeSigma
  • Modulatory effects of a tannin fraction isolated from Terminalia arjuna on the genotoxicity of mutagens in Salmonella typhimurium.
Modulatory effects of a tannin fraction isolated from Terminalia arjuna on the genotoxicity of mutagens in Salmonella typhimurium.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2000-10-18)
S J Kaur, I S Grover, S Kumar
ABSTRACT
A fraction isolated from Terminalia arjuna was studied for its antimutagenic effect against 4-nitro-o-phenylenediamine (NPD) in TA98, sodium azide in TA100 and 2-aminofluorene (2AF, S9-dependent), a promutagen, in both TA98 and TA 100 tester strains of Salmonella typhimurium using the Ames assay. The fraction inhibited the mutagenicity of 2AF very significantly in both strains while the revertant colonies induced by NPD and sodium azide were reduced moderately. 1H-NMR, 13C-NMR, IR and UV-spectroscopic data of the fraction revealed it to be tannin in nature.
Sigma-Aldrich
4-Nitro-o-phenylenediamine, 98%