MilliporeSigma
  • Dipeptide binding to the extended active site of the Streptomyces R61 D-alanyl-D-alanine-peptidase: the path to a specific substrate.
Dipeptide binding to the extended active site of the Streptomyces R61 D-alanyl-D-alanine-peptidase: the path to a specific substrate.
Biochemistry (2000-10-04)
J W Anderson, R F Pratt
ABSTRACT
Bacterial cell walls are cross-linked in the final step of biosynthesis by specific D-alanyl-D-alanine(DD)-peptidases/transpeptidases. The natural substrates of these enzymes should therefore be segments of peptidoglycan, but high specificity for such structures has yet to be demonstrated. The binding of dipeptides to the extended substrate binding site of the Streptomyces R61 DD-peptidase has been studied by means of a fluorescent beta-lactam probe. It was found that dipeptides of structure Gly-L-Xaa have affinity for a subsite adjacent to the beta-lactam binding site. Hydrophobic peptides such as Gly-L-Met and Gly-L-aminocaprylic acid had the greatest affinity for this site, with dissociation constants in each case of 0.19 mM. A combination of this motif with the C-terminal D-alanyl-D-alanine moiety required of a DD-peptidase substrate yielded a new substrate, glycyl-L-alpha-amino-epsilon-pimelyl-D-alanyl-D-alanine. Steady-state kinetic measurements established this compound as the most specific peptide substrate yet discovered for a DD-peptidase by at least 3 orders of magnitude (k(cat) = 69 s(-1), K(m) = 7.9 microM, k(cat)/K(m) = 8.7 x 10(6) s(-1) M(-1)); acylation was rate-determining at saturation. This substrate, presumably not coincidentally, contains the acyl donor and acceptor moieties, appropriately separated, of the Streptomyces peptidoglycan structure. This general method of approach should be of value in the search for specific substrates and inhibitors (antibiotics) of other DD-peptidases.
Sigma-Aldrich
N-Dodecyl-N,N-dimethyl-3-ammonio-1-propanesulfonate, ≥97.0% (dried material, CHN)
Sigma-Aldrich
N-Dodecyl-N,N-dimethyl-3-ammonio-1-propanesulfonate