MilliporeSigma
  • The selective dopamine D4 receptor antagonist, PNU-101387G, prevents stress-induced cognitive deficits in monkeys.
The selective dopamine D4 receptor antagonist, PNU-101387G, prevents stress-induced cognitive deficits in monkeys.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology (2000-09-16)
A F Arnsten, B Murphy, K Merchant
ABSTRACT
Stress exposure impairs the cognitive functioning of the prefrontal cortex (PFC). Previous research has examined the dopamine (DA) D1 receptor mechanisms underlying this response. The current study performed a preliminary examination of the role of D4 receptor mechanisms by determining whether the selective D4 receptor antagonist, PNU-101387G, could prevent stress-induced working memory deficits in monkeys. Animals were tested on the delayed response task following treatment with PNU-101387G (0 or 0.1-0.8 mg/kg, 60-min pretreatment), and the pharmacological stressor, FG7142 (0 or 0.2 mg/kg, 30-min pretreatment). FG7142 significantly impaired delayed response performance relative to vehicle; PNU-101387G pretreatment produced a dose-related reversal of the FG7142 response. PNU-101387G had no significant effects on its own, but there were trends toward improvement at low doses and impairment at higher doses. Further studies in a larger number of animals appear warranted. These preliminary findings suggest that D4 receptor mechanisms contribute to stress-induced cognitive dysfunction.
Sigma-Aldrich
Sonepiprazole hydrate, ≥98% (HPLC)