MilliporeSigma
  • Angiotensin-converting enzyme insertion/deletion polymorphism modulates the human in vivo metabolism of bradykinin.
Angiotensin-converting enzyme insertion/deletion polymorphism modulates the human in vivo metabolism of bradykinin.
Circulation (2000-08-23)
L J Murphey, J V Gainer, D E Vaughan, N J Brown
ABSTRACT
Bradykinin is a cardioprotective peptide metabolized by the angiotensin-converting enzyme (ACE). An insertion/deletion (I/D) polymorphism in the ACE gene determines plasma ACE levels. The D allele is associated with cardiovascular disease, which may relate to enhanced angiotensin II production or to increased bradykinin degradation to the inactive metabolite bradykinin 1-5 (BK1-5). Therefore, we determined the effect of the ACE I/D polymorphism on human bradykinin metabolism in vivo. Bradykinin (400 ng/min) was infused into the brachial artery of volunteers with ACE I/I, I/D, or D/D genotypes (n=9 each). The bradykinin and BK1-5 levels in forearm venous return were quantified by liquid chromatography-mass spectroscopy. Plasma ACE activity was highest in those with the D/D genotype (36.8+/-6.2 U/mL), intermediate in those with the I/D genotype (25.3+/-3.3 U/mL), and lowest in those with the I/I genotype (20.3+/-2.3 U/mL; P=0.017 for effect of number of D alleles). Bradykinin concentrations were 726+/-242, 469+/-50, and 545+/-104 fmol/mL in I/I, I/D, and D/D subjects, respectively (P>0. 10). Significant correlations existed between the number of D alleles and BK1-5 concentrations (1113+/-290, 1520+/-318, and 1887+/-388 fmol/mL in the I/I, I/D, and D/D groups, respectively; P=0.027) and the ratio of BK1-5 to bradykinin (1.87+/-0.35, 3.09+/-0. 40, and 4.31+/-0.97 in the I/I, I/D, and D/D volunteers, respectively; P=0.010). The venous blood BK1-5:bradykinin ratio correlated with plasma ACE activity (r(2)=0.16, P=0.039), and total kinin concentration correlated with net tissue plasminogen activator release across the forearm (r(2)=0.20, P=0.027). The ACE D allele has a significant effect on the in vivo degradation of bradykinin in humans. The ratio of BK1-5:bradykinin may serve as a marker for tissue ACE activity.
Sigma-Aldrich
Bradykinin Fragment 1-5, ≥97% (HPLC)