MilliporeSigma
[Hydrolysis of primary amide groups in Asn/Gln-containing peptides].
Bioorganicheskaia khimiia (2000-08-03)
V V Onoprienko, E A Elin, A I Miroshnikov
ABSTRACT
Peptides Boc-Ala-Asn/Gln-OH and Boc-Asn/Gln-Ala-OH were saponified with barium hydroxide to corresponding Asp/Glu-containing peptides. Under the conditions of saponification, Boc-Asn-Ala-OH additionally afforded Boc-Asp-OH, isopeptide Boc-Asp(Ala)-OH, and Boc-NHSuc > Ala-OH, with the third being the key intermediate in these transformations. Boc-Asp(OMe)-Ala-OMe underwent similar transformations under treatment with diazomethane or triethylamine. Saponification with barium hydroxide was accompanied by a high epimerization of N-terminal amino acid residues, whereas the products of the diazomethane treatment of Boc-Asp(OMe)-Ala-OMe had a low degree of epimerization.
Sigma-Aldrich
Barium hydroxide, technical grade, ~95%
Sigma-Aldrich
Barium hydroxide solution, 0.3 N
Sigma-Aldrich
Barium hydroxide octahydrate, ≥98%
Sigma-Aldrich
Barium hydroxide octahydrate, ACS reagent, ≥98%
Sigma-Aldrich
Barium hydroxide monohydrate, 98%