MilliporeSigma
Cytotoxic neolignans from the stem wood of Machilus obovatifolia.
Planta medica (2000-07-26)
I L Tsai, J H Chen, C Y Duh, I S Chen
ABSTRACT
Four new cytotoxic neolignans, machilusol A (1), machilusol C (3), machilusol D (4), machilusol E (5) as well as two new inactive neolignans, machilusol B (2) and machilusol F (6), were isolated from the stem wood of Machilus obovatifolia. All structures were identified by means of spectroscopic analysis.