MilliporeSigma
  • Micelle-mediated extraction for concentrating hydrophobic organic compounds.
Micelle-mediated extraction for concentrating hydrophobic organic compounds.
Journal of chromatography. A (2000-07-13)
T Saitoh, T Matsudo, C Matsubara
ABSTRACT
An extraction method based on polymer-induced phase separation of aqueous micellar solutions of octyl-beta-D-thioglucoside (OTG) was assessed for concentrating hydrophobic analytes. Various hydrophobic compounds such as polycyclicaromatic hydrocarbons, alkylbenzenes, alkylphenols, chlorobenzenes, chlorophenols, phthalic esters, pesticides, and steroid hormones could be efficiently concentrated into a small volume of surfactant-rich phase, while hydrophilic matrix components remained in the bulk aqueous phase. The surfactant-rich phase containing concentrated OTG could be directly introduced into the hygro-organic mobile phase of high-performance liquid chromatography with ultra-violet photometric detection. The application of this method greatly enhanced the signal intensity in the chromatogram while reducing the interference of matrix components.
Sigma-Aldrich
Octyl β-D-1-thioglucopyranoside, ≥98.0% (GC)