MilliporeSigma
  • Bromoacetophenone-based photonucleases: photoinduced cleavage of DNA by 4'-bromoacetophenone-pyrrolecarboxamide conjugates.
Bromoacetophenone-based photonucleases: photoinduced cleavage of DNA by 4'-bromoacetophenone-pyrrolecarboxamide conjugates.
Organic letters (2000-06-03)
P A Wender, R Jeon
ABSTRACT
[formula: see text] 4'-Bromoacetophenone derivatives which upon excitation can generate monophenyl radicals capable of hydrogen atom abstraction were investigated as photoinducible DNA cleaving agents. Pyrrolecarboxamide-conjugated 4'-bromoacetophenones were synthesized, and their DNA cleaving activities and sequence selectivities were determined.
Product Description
Supelco
2-Bromoacetophenone, derivatization grade (GC derivatization), LiChropur, ≥99.0%
Sigma-Aldrich
2-Bromoacetophenone, 98%