MilliporeSigma
  • Guest dechlorination and covalent capture following photoexcitation of inclusion complexes in water.
Guest dechlorination and covalent capture following photoexcitation of inclusion complexes in water.
Chemosphere (2000-05-02)
D A Williamson, A M Barenberg, C A Coleman, D R Benson
ABSTRACT
Photoexcitation of complexes between cyclophane 1 and 1- or 2-chloronaphthalene in aqueous solution leads to rapid dechlorination of the guest, a reaction driven by electron transfer from host to excited guest. The main photoproducts contain a naphthyl group covalently attached to the host framework. The results may lead to new approaches for remediating water contaminated with chlorinated aromatic compounds.
Sigma-Aldrich
1-Chloronaphthalene, technical, ≥85% (GC)
Sigma-Aldrich
1-Chloronaphthalene, technical grade