MilliporeSigma
  • Acute metobromuron poisoning with severe associated methemoglobinemia. Identification of four metabolites in plasma and urine by LC-DAD, LC-ESI-MS, and LC-ESI-MS-MS.
Acute metobromuron poisoning with severe associated methemoglobinemia. Identification of four metabolites in plasma and urine by LC-DAD, LC-ESI-MS, and LC-ESI-MS-MS.
Journal of analytical toxicology (2000-04-25)
A Turcant, A Cailleux, A Le Bouil, P Allain, P Harry, A Renault
ABSTRACT
A case of self poisoning with metobromuron, a urea derivative used as a herbicide, is reported. Severe methemoglobinemia observed at the admission (80%) disappeared only at day 11, and hemolysis appeared at day 4 and decreased slowly to day 12. Metobromuron was analyzed by liquid chromatography with diode-array detection. Initial plasma concentration and elimination half-life were 4.9 mg/L and 5 h, respectively. Several metabolites were also detected, and four of those were identified by liquid chromatography-electrospray mass spectrometry. Normetobromuron, bromophenylurea, and bromoacetanilide were detected in plasma, but only N-methyl bromophenylurea was detected in urine. Bromoacetanilide probably results from acetylation of the intermediate bromoaniline. Methemoglobinemia could result from metabolization of metobromuron to bromoaniline and bromoacetanilide.
Sigma-Aldrich
4-Bromoaniline, ≥99.0% (GC)
Sigma-Aldrich
4-Bromoaniline, 97%