MilliporeSigma
  • The preservatives ethyl-, propyl- and butylparaben are oestrogenic in an in vivo fish assay.
The preservatives ethyl-, propyl- and butylparaben are oestrogenic in an in vivo fish assay.
Pharmacology & toxicology (2001-02-07)
K L Pedersen, S N Pedersen, L B Christiansen, B Korsgaard, P Bjerregaard
ABSTRACT
The widely used phenolic preservatives ethylparaben, propylparaben, butylparaben and their common metabolite p-hydroxybenzoic acid were tested for their ability to evoke an oestrogenic response in vivo. Yolk protein induction in sexually immature rainbow trout was used as an oestrogen-specific endpoint after repeated injections of the compounds. All tested parabens were oestrogenic in doses between 100 and 300 mg/kg, while the metabolite showed no activity. Ethylparaben was found to be approximately sixty times weaker than propyl- and butylparaben which had oestrogenic potencies comparable to those previously found for bisphenol A.
Product Description
Ethyl parahydroxybenzoate, European Pharmacopoeia (EP) Reference Standard
Supelco
Ethylparaben, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethyl 4-hydroxybenzoate, ReagentPlus®, 99%