MilliporeSigma
  • Vicinal diketone formation in yogurt: (13)C precursors and effect of branched-chain amino acids.
Vicinal diketone formation in yogurt: (13)C precursors and effect of branched-chain amino acids.
Journal of agricultural and food chemistry (2000-03-22)
A Ott, J E Germond, A Chaintreau
ABSTRACT
Addition of branched-chain amino acids (BCAA) or an inhibitor of the BCAA biochemical pathways during fermentation of milk with a lac(-) mutant of Lactobacillus delbrueckii subsp. bulgaricus and Streptococcus thermophilus strongly influenced the formation of two aroma-impact compounds, 2,3-butanedione and 2,3-pentanedione, as well as their direct precursors 2-acetolactate and 2-acetohydroxybutyrate. This suggests a connection between vicinal diketone formation and BCAA biosynthesis in yogurt bacteria. A recently developed static-and-trapped headspace technique combined with gas chromatography-mass spectrometry demonstrated incorporation of (13)C from [U-(13)C(6)]-D-glucose and [U-(13)C(4)]-L-threonine into both vicinal diketones. For 2,3-butanedione, glucose is the major precursor via pyruvate and activated acetaldehyde. For 2, 3-pentanedione, L-threonine is a precursor via 2-ketobutyrate, but glucose is the major contributor via activated acetaldehyde and, possibly, also via 2-ketobutyrate, which is a degradation product of 3-methylaspartate, an intermediate in glutamate synthesis.
Sigma-Aldrich
2,3-Pentanedione, 97%
Sigma-Aldrich
2,3-Pentanedione, ≥96%, FCC, FG