MilliporeSigma
  • Comparison of the mutagenic specificity induced by four nitro-group-containing aromatic amines in Salmonella typhimurium his genes.
Comparison of the mutagenic specificity induced by four nitro-group-containing aromatic amines in Salmonella typhimurium his genes.
Mutation research (2000-03-10)
K Chung, T J Hughes, L D Claxton
ABSTRACT
Four nitrated aromatic amines (2-nitro-p-phenylenediamine [2NPD], 3-nitro-o-phenylenediamine [3NPD], 4-nitro-o-phenylenediamine [4NPD] and 4,4'-dinitro-2-biphenylamine [DNBA]) are direct-acting mutagens in Salmonella typhimurium strain TA100. These compounds were tested further using the Xenometrix strains of S. typhimurium: TA7001, TA7002, TA7003, TA7004, TA7005, and TA7006, with and without S9 mix in the plate incorporation assay. The direct-acting mutagenicity of 2NPD, 4NPD, and DNBA was detected with TA7002, TA7004 and TA7005. 2NPD and DNBA showed some activity in TA7006; DNBA also showed some activity in TA7003. Mutagenicity was generally decreased in these strains when S9 was added. 3NPD was mutagenic in TA7004 without S9 and in TA7005 with and without S9. These data suggest that 2NPD, 4NPD and DNBA induced TA-->AT and CG-->AT transversions as well as GC-->AT transitions in the his gene. 3NPD induced CG-->AT transversions and GC-->AT transitions. 2NPD and DNBA also induced a small portion of CG-->GC transversions.
Sigma-Aldrich
4-Nitro-o-phenylenediamine, 98%
Sigma-Aldrich
2-Nitro-1,4-phenylenediamine, 95%