MilliporeSigma
  • Exposure to airborne dust contaminated with pesticide in the Aral Sea region.
Exposure to airborne dust contaminated with pesticide in the Aral Sea region.
Lancet (London, England) (2000-03-04)
S L O'Hara, G F Wiggs, B Mamedov, G Davidson, R B Hubbard
ABSTRACT
The Aral Sea region is one of the world's foremost ecological disaster zones and there is increasing local concern for the health of millions of people living in this region. We have found that dust deposition rates across eastern Turkmenistan are among the highest in the world and that the dust is contaminated with pesticide.
Product Description
Supelco
Phosalone, PESTANAL®, analytical standard