MilliporeSigma
  • Dysregulation of adrenal 11 beta-hydroxylase activity in hypertensive subjects: usefulness of the ACTH 1-17 stimulation test.
Dysregulation of adrenal 11 beta-hydroxylase activity in hypertensive subjects: usefulness of the ACTH 1-17 stimulation test.
Nutrition, metabolism, and cardiovascular diseases : NMCD (1999-12-30)
R Valentino, A P Tommaselli, S Savastano, M T Scarpitta, M Dorato, A Luciano, E Calvanese, G Lombardi
ABSTRACT
The aim of this study was to determine the validity of our previous hypothesis of adrenal 11 beta-hydroxylase (11-OH) dysregulation in "essential" low-renin hypertension. A comparison was made between 30 hypertensive patients and 30 age-matched controls (NC) in basal conditions and after ACTH stimulation (ACTH 1-17) test. The 11-deoxycortisol (S) and deoxycorticosterone (DOC) integrated areas under the curve (AUCs) of stimulus were significantly higher in the hypertensives (p < 0.001) and pointed to adrenal 11-OH dysregulation. The ACTH 1-17 test detects impairment of 11-OH activity of probable genetic origin. The relative mineralocorticoid excess thus provoked could be an additional cause of "essential" low-renin hypertension.
Sigma-Aldrich
Adrenocorticotropic Hormone Fragment 1-17 human, rat, ≥97% (HPLC)