MilliporeSigma
  • Continuous degradation of mixtures of 4-nitrobenzoate and 4-aminobenzoate by immobilized cells of Burkholderia cepacia strain PB4.
Continuous degradation of mixtures of 4-nitrobenzoate and 4-aminobenzoate by immobilized cells of Burkholderia cepacia strain PB4.
Applied microbiology and biotechnology (1999-12-28)
C M Peres, B Van Aken, H Naveau, S N Agathos
ABSTRACT
Although isolated on 4-aminobenzoate, Burkholderia cepacia strain PB4 is also able to grow on 4-nitrobenzoate. Degradation of an equimolar mixture of the nitroaromatic compound 4-nitrobenzoate and its corresponding aminoaromatic derivative 4-aminobenzoate by this strain was investigated. Batch experiments showed that, irrespective of preculturing conditions, both compounds were degraded simultaneously. The mixture-degrading ability of B. cepacia strain PB4 was subsequently tested in continuous packed bed reactors (PBR) with the strain immobilized on Celite grade R-633 or R-635. Higher degradation rates were achieved with the larger particles of Celite R-635. Maximum simultaneous degradation rates per liter of packed bed of 0.925 mmol 1(-1) h(-1) 4-nitrobenzoate and 4-aminobenzoate were obtained for an applied loading rate of the same value (0.925 mmol 1(-1) h(-1) of each compound). Even when the applied load was not removed in its entirety, neither of the two compounds was degraded preferentially but a percentage of both of them was mineralized. The present study shows the possibility for a pure strain to biodegrade not only a nitroaromatic compound (4-nitrobenzoate) but also its corresponding amino derivative (4-aminobenzoate) continuously and simultaneously.
Sigma-Aldrich
4-Nitrobenzoic acid, 98%
Sigma-Aldrich
4-Nitrobenzoic acid, purum, ≥98.0% (HPLC)