MilliporeSigma
  • Comparison of in vitro and ex vivo antiplatelet effects of 1,2-benzisothiazolin-3-one and its 2-amino derivative.
Comparison of in vitro and ex vivo antiplatelet effects of 1,2-benzisothiazolin-3-one and its 2-amino derivative.
Arzneimittel-Forschung (1999-12-22)
P Vicini, C Manotti, A Caretta, L Amoretti
ABSTRACT
The in vitro and ex vivo antiplatelet effects of 2-amino-1,2-benzisothiazolin-3-one (1) are compared with those of its parent compound 1,2-benzisothiazolin-3-one (2) and with acetylsalicylic acid (ASA) against different agonists. 2-Amino-1,2-benzisothiazolin-3-one inhibits adenosine diphosphate (ADP)-, arachidonic acid (AA)- and collagen-induced human platelet aggregation in vitro, with IC50 values of 8.90 x 10(-5), 1.50 x 10(-6) and 5.11 x 10(-8) mol/l, respectively. The strong inhibitory activity is significant not only for collagen but also for AA-induced aggregation. The same compound inhibits ex vivo collagen- and particularly AA-induced rabbit platelet aggregation at the tested dose of 10 mg/kg i.m. In view of the potential use of 2-amino-1,2-benzisothiazolin-3-one as antithrombotic agent, the log P values for both 1,2-benzisothiazolin-3-one derivatives 1 and 2 are determined, to gain an understanding of the significance of the 2-amino group in the 1,2-benzisothiazolin-3-one moiety with respect to the biological activity under study.
Sigma-Aldrich
1,2-Benzisothiazol-3(2H)-one, 97%
Supelco
1,2-Benzisothiazol-3(2H)-one, analytical standard