MilliporeSigma
  • Regiospecific internal desaturation of aliphatic compounds by a mutant Rhodococcus strain.
Regiospecific internal desaturation of aliphatic compounds by a mutant Rhodococcus strain.
Applied and environmental microbiology (1999-12-03)
K Koike, K Ara, S Adachi, H Takigawa, H Mori, S Inoue, Y Kimura, S Ito
ABSTRACT
A mutant Rhodococcus strain lacking the ability to utilize 1-chlorohexadecane was found to cis-desaturate aliphatic compounds, such as 1-chlorohexadecane, n-hexadecane, and heptadecanonitrile, yielding corresponding products with a double bond mainly at the ninth carbon from the terminal methyl groups. A new oxidative pathway involving the cis-desaturation step was suggested for alkane utilization by Rhodococcus spp.
Sigma-Aldrich
1-Chlorohexadecane, 95%