MilliporeSigma
  • Characterization of volatile compounds from the reaction of 3-hydroxy-2-butanone and ammonium sulfide model system.
Characterization of volatile compounds from the reaction of 3-hydroxy-2-butanone and ammonium sulfide model system.
Journal of agricultural and food chemistry (1999-11-24)
J Xi, T C Huang, C T Ho
ABSTRACT
The reactions between 3-hydroxy-2-butanone and ammoniun sulfide at 25, 50, 75, 100, 125, and 150 degrees C were studied. Four well-known flavor compounds, 2,4,5-trimethyloxazole, 2,4, 5-trimethyl-3-oxazoline, 2,4,5-trimethylthiazole, and 2,4, 5-trimethyl-3-thiazoline, were identified. Another four interesting intermediate compounds, 2-(1-hydroxyethyl)-2,4, 5-trimethyl-3-oxazoline, 2-(1-mercaptoethyl)-2,4, 5-trimethyl-3-oxazoline, 2-(1-hydroxyethyl)-2,4, 5-trimethyl-3-thiazoline, and 2-(1-mercaptoethyl)-2,4, 5-trimethyl-3-thiazoline, were also identified by GC-EIMS and GC-CIMS. All these intermediate compounds were formed at 25 degrees C. On the other hand, tetramethylpyrazine was the major product with a reaction temperature higher than 100 degrees C.
Sigma-Aldrich
Ammonium sulfide solution, 20 wt. % in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
Ammonium sulfide solution, 20% in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
Ammonium sulfide solution, 40-48 wt. % in H2O
Pricing and availability is not currently available.