MilliporeSigma
  • Unhydrolyzable analogues of adenosine 3':5'-monophosphate demonstrating growth inhibition and differentiation in human cancer cells.
Unhydrolyzable analogues of adenosine 3':5'-monophosphate demonstrating growth inhibition and differentiation in human cancer cells.
Cancer research (1992-05-01)
H Yokozaki, G Tortora, S Pepe, E Maronde, H G Genieser, B Jastorff, Y S Cho-Chung
ABSTRACT
A set of adenosine 3':5'-monophosphate (cAMP) analogues that combine exocyclic sulfur substitutions in the equatorial (Rp) or the axial (Sp) position of the cyclophosphate ring with modifications in the adenine base of cAMP were tested for their effect on the growth of HL-60 human promyelocytic leukemia cells and LS-174T human colon carcinoma cells. Both diasteromeres of the phosphorothioate derivatives were growth inhibitory, exhibiting a concentration inhibiting 50% of cell proliferation of 3-100 microM. Among the analogues tested, Rp-8-Cl-cAMPS and Sp-8-Br-cAMPS were the two most potent. Rp-8-Cl-cAMPS was 5- to 10-fold less potent than 8-Cl-cAMP while Sp-8-Br-cAMPS was approximately 6-fold more potent than 8-Br-cAMP. The growth inhibition was not due to a block in a specific phase of the cell cycle or due to cytotoxicity. Rp-8-Cl-cAMPS enhanced its growth-inhibitory effect when added together with 8-Cl-cAMP and increased differentiation in combination with N6-benzyl-cAMP. The binding kinetics data showed that these Sp and Rp modifications brought about a greater decrease in affinity for Site B than for Site A of RI (the regulatory subunit of type I cAMP-dependent protein kinase) and a substantial decrease of affinity for Site A of RII (the regulatory subunit of type II protein kinase) but only a small decrease in affinity for Site B of RII, indicating the importance of the Site B binding of RII in the growth-inhibitory effect. These results show that the phosphorothioate analogues of cAMP are useful tools to investigate the mechanism of action of cAMP in growth control and differentiation and may have practical implication in the suppression of malignancy.