MilliporeSigma
  • A continuous assay for the spectrophotometric analysis of sulfotransferases using aryl sulfotransferase IV.
A continuous assay for the spectrophotometric analysis of sulfotransferases using aryl sulfotransferase IV.
Analytical biochemistry (1999-10-21)
M D Burkart, C H Wong
ABSTRACT
We have developed a continuous spectrophotometric coupled-enzyme assay for sulfotransferase activity. This assay is based on the regeneration of 3'-phosphoadenosine-5'-phosphosulfate (PAPS) from the desulfated 3'-phosphoadenosine-5'-phosphate (PAP) by a recombinant aryl sulfotransferase using p-nitrophenyl sulfate as the sulfate donor and visible spectrophotometric indicator of enzyme turnover. Here recombinant rat aryl sulfotransferase IV (AST-IV) is expressed, resolved to the pure beta-form during purification, and utilized for the regeneration. The activity of betaAST-IV to catalyze the synthesis of PAPS from PAP and p-nitrophenyl sulfate is demonstrated via capillary zone electrophoresis, and the kinetics of this reverse-physiological reaction are calculated. betaAST-IV is then applied to the coupled enzyme system, where the steady-state activity of the commercially available Nod factor sulfotransferase is verified with an enzyme concentration study and substrate-specificity assays of N-chitoses. The potential applications of this assay include rapid kinetic determinations for carbohydrate and protein sulfotransferases, high-throughput screening of potential sulfotransferase substrates and inhibitors, and biomedical screening of blood samples and other tissues for specific sulfotransferase enzyme activity and substrate concentration.
Sigma-Aldrich
Potassium 4-nitrophenyl sulfate, sulfatase substrate