MilliporeSigma
  • The reactivity of the 2-deoxyribonolactone lesion in single-stranded DNA and its implication in reaction mechanisms of DNA damage and repair.
The reactivity of the 2-deoxyribonolactone lesion in single-stranded DNA and its implication in reaction mechanisms of DNA damage and repair.
Nucleic acids research (1999-09-11)
J T Hwang, K A Tallman, M M Greenberg
ABSTRACT
The formal C1'-oxidation product, 2-deoxyribonolactone, is formed as a result of DNA damage induced via a variety of agents, including gamma-radiolysis and the enediyne antitumor antibiotics. This alkaline labile lesion may also be an intermediate during DNA damage induced by copper-phenanthroline. Oligo-nucleotides containing this lesion at a defined site were formed via aerobic photolysis of oligonucleotides containing a photolabile ketone, and were characterized by gel electrophoresis and electrospray mass spectrometry (ESI-MS). Treatment of oligo-nucleotides containing the lesion with secondary amines produces strand breaks consisting of 3'-phosphate termini, and products which migrate more slowly in polyacrylamide gels. MALDI-TOF mass spectrometry analysis indicates that the slower moving products are formal adducts of the beta-elimination product resulting from 2-deoxyribonolactone and one molecule of amine. The addition of beta-mercapto-ethanol to the reaction mixture produces thiol adducts as well. The stability of these adducts suggests that they cannot be the labile species characterized by gel electrophoresis in copper-phenanthroline-mediated strand scission. The characterization of these adducts by mass spectrometry also provides, by analogy, affirmation of proposals regarding the reactivity of nucleophiles with the beta-elimination product of abasic sites. Finally, the effects of this lesion and the various adducts on DNA repair enzymes are unknown, but their facile generation from oligonucleotides containing a photolabile ketone suggests that such issues could be addressed.
Sigma-Aldrich
N,N′-Dimethylethylenediamine, 85%
Sigma-Aldrich
N,N′-Dimethylethylenediamine, 98%