MilliporeSigma
  • Metabolic activation and inactivation of metanil yellow and orange II in the Salmonella typhimurium his-reversion assay.
Metabolic activation and inactivation of metanil yellow and orange II in the Salmonella typhimurium his-reversion assay.
Journal of food protection (1996-03-01)
P B Rastogi, R E Levin
ABSTRACT
Among a variety of experimental protocols used, the combined use of 0.5% dextrose in bottom agar and 1 micromol of flavin mononucleotide (FMN) in preincubation mixtures without fraction S9 mix resulted in the highest level of induced Salmonella typhimurium his reversions with both dyes metanil yellow and orange 11 with strain TA100. Strain TA98 yielded notably lower levels of reversions under the same conditions. The presence of uninduced hamster liver S9 fraction resulted in a weak mutagenic response while Aroclor 1254-induced rat liver S9 fraction resulted in the complete absence of mutagenicity with both strains and with both dyes.
Product Description
Supelco
Metanil Yellow, analytical standard
Sigma-Aldrich
Metanil Yellow, Dye content 70 %