MilliporeSigma
  • Anion channel blockers differentially affect T-type Ca(2+) currents of mouse spermatogenic cells, alpha1E currents expressed in Xenopus oocytes and the sperm acrosome reaction.
Anion channel blockers differentially affect T-type Ca(2+) currents of mouse spermatogenic cells, alpha1E currents expressed in Xenopus oocytes and the sperm acrosome reaction.
Developmental genetics (1999-08-10)
F Espinosa, I López-González, C J Serrano, G Gasque, J L de la Vega-Beltrán, C L Treviño, A Darszon
ABSTRACT
The direct electrophysiological characterization of sperm Ca(2+) channels has been precluded by their small size and flat shape. An alternative to study these channels is to use spermatogenic cells, the progenitors of sperm, which are larger and easier to patch-clamp. In mouse and rat, the only voltage-dependent Ca(2+) currents displayed by these cells are of the T type. Because compounds that block these currents inhibit the zona pellucida-induced Ca(2+) uptake and the sperm acrosome reaction (AR) at similar concentrations, it is likely that they are fundamental for this process. Recent single channel recordings in mouse sperm demonstrated the presence of a Cl(-) channel. This channel and the zona pellucida (ZP)-induced AR were inhibited by niflumic acid (NA), an anion channel blocker [Espinosa et al. (1998): FEBS Lett 426:47-51]. Because NA and other anion channel blockers modulate cationic channels as well, it became important to determine whether they affect the T-type Ca(2+) currents of spermatogenic cells. These currents were blocked in a voltage-dependent manner by NA, 1, 9-dideoxyforskolin (DDF), and 5-nitro-2-(3-phenylpropylamine)benzoic acid (NPPB). The IC(50) values at -20 mV were 43 microM for NA, 28 microM for DDF, and 15 microM for NPPB. Moreover, DDF partially inhibited the ZP-induced AR (40% at 1 microM) and NPPB displayed an IC(50) value of 6 microM for this reaction. These results suggest that NA and DDF do not inhibit the ZP-induced AR by blocking T-type Ca(2+) currents, while NPPB may do so. Interestingly 200 microM NA was basically unable to inhibit alpha1E Ca(2+) channels expressed in Xenopus oocytes, questioning that this alpha subunit codes for the T-type Ca(2+) channels present in spermatogenic cells. Evidence for the presence of alpha1C, alpha1G, and alpha1H in mouse pachytene spematocytes and in round and condensing spermatids is presented.
Sigma-Aldrich
1,9-Dideoxyforskolin from Coleus forskohlii, ≥97%, solid