MilliporeSigma
  • Proton NMR relaxation studies of solid tyrosine derivatives and their mixtures with L-leucinamide.
Proton NMR relaxation studies of solid tyrosine derivatives and their mixtures with L-leucinamide.
Solid state nuclear magnetic resonance (1999-07-17)
Y L Wang, P S Belton, H R Tang
ABSTRACT
Proton NMR relaxation time measurements were carried out on solid tyrosine derivatives: acetyl-L-tyrosine ethyl ester (Ac-Tyroet), N-carbobenzyloxy-L-tyrosine ethyl ester (CBZ-Tyroet), N-trifluoroacetyl-L-tyrosine ethyl ester (TFAc-Tyroet) and their mixtures with L-leucinamide. It was found that spin-lattice relaxation was driven mainly by methyl group rotation it low temperature for the pure solids and the mixtures. Benzene ring flipping motion and a third motion (possibly whole molecule tumbling) were found to be responsible for relaxing Ac-Tyroet and CBZ-Tyroet at high temperature. However, these motions were highly hindered in TFAc-Tyroet. Molecular tumbling motion was detected in the supercooled liquid mixtures of L-leuNH/CBZ-Tyroet and L-leuNH/Ac-Tyroet, while this motion is absent in the mixture L-leuNH/TFAc-Tyroet. The hindered motion in TFAc-Tyroet may be one of the factors affecting its ability to form a supercooled liquid with L-leucinamide.
Sigma-Aldrich
L-Leucinamide hydrochloride, 99%