MilliporeSigma
  • Crosslinking and modification of dermal sheep collagen using 1, 4-butanediol diglycidyl ether.
Crosslinking and modification of dermal sheep collagen using 1, 4-butanediol diglycidyl ether.
Journal of biomedical materials research (1999-07-09)
R Zeeman, P J Dijkstra, P B van Wachem, M J van Luyn, M Hendriks, P T Cahalan, J Feijen
ABSTRACT
Crosslinking of dermal sheep collagen (DSC) was accomplished using 1, 4-butanediol diglycidyl ether (BDDGE). At pH values > 8.0, epoxide groups of BDDGE will react with amine groups of collagen. The effects of BDDGE concentration, pH, time, and temperature were studied. Utilization of a 4-wt % BDDGE instead of 1-wt % resulted in a faster reaction. Whereas similar values of shrinkage temperature were obtained, fewer primary amine groups had reacted at a lower BDDGE concentration, which implies that the crosslinking reaction had a higher efficacy. An increase in pH from 8.5 to 10.5 resulted in a faster reaction but reduced crosslink efficacy. Furthermore, an increase in reaction temperature accelerated the reaction without changing the crosslink efficacy. Crosslinking under acidic conditions (pH < 6.0) evoked a reaction between epoxide groups and carboxylic acid groups of collagen. Additional studies showed that no oligomeric crosslinks could be formed. However, hydrolysis of the epoxide groups played a role in the crosslink mechanism especially under acidic reaction conditions. The macroscopic properties of these materials were dependent on the crosslinking method. Whereas a flexible and soft tissue was found if crosslinking was performed at pH < 6.0, a stiff sponge was obtained under alkaline conditions. Reaction of DSC with a monofunctional compound (glycidyl isopropyl ether) led to comparable trends in reaction rate and in similar macroscopical differences in materials as observed with BDDGE.
Sigma-Aldrich
1,4-Butanediol diglycidyl ether, technical grade, 60%
Sigma-Aldrich
1,4-Butanediol diglycidyl ether, ≥95%