MilliporeSigma
  • Characterization of the myosin light chain kinase from smooth muscle as an actin-binding protein that assembles actin filaments in vitro.
Characterization of the myosin light chain kinase from smooth muscle as an actin-binding protein that assembles actin filaments in vitro.
Biochimica et biophysica acta (1999-05-08)
K Hayakawa, T Okagaki, L H Ye, K Samizo, S Higashi-Fujime, T Takagi, K Kohama
ABSTRACT
In addition to its kinase activity, myosin light chain kinase has an actin-binding activity, which results in bundling of actin filaments [Hayakawa et al., Biochem. Biophys. Res. Commun. 199, 786-791, 1994]. There are two actin-binding sites on the kinase: calcium- and calmodulin-sensitive and insensitive sites [Ye et al., J. Biol. Chem. 272, 32182-32189, 1997]. The calcium/calmodulin-sensitive, actin-binding site is located at Asp2-Pro41 and the insensitive site is at Ser138-Met213. The cyanogen bromide fragment, consisting of Asp2-Met213, is furnished with both sites and is the actin-binding core of myosin light chain kinase. Cross-linking between the two sites assembles actin filaments into bundles. Breaking of actin-binding at the calcium/calmodulin-sensitive site by calcium/calmodulin disassembles the bundles.
Sigma-Aldrich
2-Nitro-5-thiocyanatobenzoic acid, powder